Learn More
Greiner Bio-One CELLCOAT™ Poly-D-Lysine Protein Coated Flasks
Click to view available options
Area di coltura:
175 cm2
Volume (sistema metrico) di lavorazione:
50 mL
650mL
Dimensione della confezione:
pezzi 40
pezzi 50
Descrizione
Poly-D-lysine (PDL) are synthetic molecules which increase cell adhesion to vessels by altering the net charge of the surface without stimulating biological activity.
- Coated with Poly-D-Lysine synthetic protein (75-150 kDa)
- For promotion of cell adhesion, proliferation and growth of transfected cell lines, neuronal cell lines, primary neurons and glial cells
- Ready-to-use products
- Includes lid with condensation rings
Specifica
Specifica
| Rivestimento superficiale | Poly-D-Lysine |
| Tipo di chiusura | Plug Seal Cap |
| Area di coltura | 175 cm2 |
| Volume (sistema metrico) di lavorazione | 650mL |