Learn More
Corning™ CellCube™ Modules, With Aseptic Connectors
Sfoglia Tutto Corning Prodotti
Click to view available options
Livelli vassoi:
10-Layer
100-Layer
25-Layer
Dimensione della confezione:
Pezzo
pezzi 2
Descrizione
- Uses a tissue culture-treated growth surface for cell attachment, and continually perfuses the cells with fresh medium for increased cell productivity
- The CellCube system provides an environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen with low differential gradients across all cells within the modules.
- The CellCube modules provide a traditional tissue culture-treated surface or Corning CellBIND™ surface for the growth of attachment dependent cells.
Specifica
Specifica
| Include | Growth Surface and Aseptic Connectors |
| N. per custodia | 2/Cs. |
| N. per confezione | 1/Pk. |
| Sterilità | Sterile |
| Area di coltura | 8500 cm² |
| Livelli vassoi | 10-Layer |