Learn More
Greiner Bio-One Matracci per colture cellulari Cellstar™
Click to view available options
Capacità (sistema metrico):
50 mL
250 ml
550 ml
650 mL
Area di coltura:
175 cm2
75 cm2
Tipo di chiusura:
Screw Cap with Vent Position
Tappo a vite per filtro rosso
Include:
Graduations; writing area
Printed and engraved graduations
Printed and engraved graduations; measuring grid
Printed graduations; labeling field on side
Printed graduations; labeling field on the side
Printed graduations; writing area
Dimensione della confezione:
pezzi 120
pezzi 200
pezzi 40
pezzi 50
Descrizione
- Improved cell adhesion through physical surface treatment
- Neck of high profile flask reduces the risk of wetting the inside of the cap with medium; provides additional protection against contamination
- Canted neck
- TC surface treatment
- Graduation on the side
- Stackable
- Measuring grid
- Red standard screw cap
- Sterile and user-friendly packaging
- Free of detectable DNase, RNase, human DNA, non-pyrogenic
- Non-Cytotoxic
Specifica
Specifica
| Colore chiusura | Red |
| Materiale chiusura | HDPE |
| Capacità (sistema metrico) | 50 mL |
| Materiale | Polystyrene |
| Include | Printed and engraved graduations |
| Stile collo | Canted |
| Rivestimento superficiale | Physical Surface Treatment |
| Graduated | Yes |
| Sterilità | Sterile |
| Tipo di chiusura | Screw Cap with Vent Position |
| Vedi altri risultati |