Learn More
Eppendorf™ CellXpert™ C170i, Segmented Inner Door, Water and Humidity Monitoring Copper Package
Click to view available options
Tipo di spina:
EU
Door Style:
Left-Sided Handle
Right-Sided Handle
Dimensione della confezione:
Pezzo
Descrizione
- Water and humidity monitoring packaging
- Copper package (chamber, shelves and water tray)
- Advanced user Interface and front USB-ports
- Fan-less design, smart racking system and innovative insulation concept
- New magnetic door locking concept for outer and inner doors
- Seamless chamber, easy disassembling
- 180°C High Temperature Disinfection (HTD) with protocol, no internal HEPA-filters
- Customizable tasks and alarms
- Standard building management system and VisioNize connected lab
- Vibration and turbulence protection
Specifica
Specifica
| Capacità (sistema metrico) | 170 L |
| Materiale camera | Acciaio inox |
| Tecnologia sensore CO2 | Sensore IR |
| Tipo di spina | EU |
| Temperature Range (Metric) | Ambient +4° to 50°C |
| Display | VisioNize™ touch interface |
| Door Style | Left-Sided Handle |
| Tecnologia sensore O2 | ZrO2 (zirconium dioxide) |
| Porte | 2 |
| Dimensioni interne (L x P x A) | 44.5 × 53.9 × 69.2 cm (17.5 × 21.2 × 27.2 in.) |
| Vedi altri risultati |