Learn More
Certified Reference Flash Point Standard - PMCC Lubricant, UKAS ISO 17034 Certified 250mL (nominal value: 190.5 °C), Paragon Scientific™
Codice prodotto. 15308528
Click to view available options
Quantità:
250 mL
Dimensione della confezione:
250mL
Codice prodotto. 15308528
Marca: Paragon Scientific CRMUPMLU
Flash Point Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Flash point according to ASTM D93.
  • Certified Reference Material for annual verification checks in accordance with ASTM D93.
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
  • Fully traceable to international standards.
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • 12 month shelf life.
  • Manufactured in the United Kingdom
Specifica
Quantità 250 mL
Nominal values quoted. Certified Values will vary from batch to batch.