missing translation for 'onlineSavingsMsg'
Learn More

CH-223191, MedChemExpress

Codice prodotto. p-200099747
Encompass_Preferred
Click to view available options
Quantità:
5 mg
10 mg
50 mg
100 mg
200 mg
Dimensione della confezione:
100mg
10mg
200mg
50mg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 16485288

Marca: Medchem Express HY12684

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

MedChemExpress CH-223191 is a potent and specific antagonist of aryl hydrocarbon receptor (AhR). CH-223191 inhibits TCDD-mediated nuclear translocation and DNA binding of AhR, and inhibits TCDD-induced luciferase activity with an IC50 of 0.03 μM.

CH-223191 is a potent and specific antagonist of aryl hydrocarbon receptor (AhR).

Specifica

Materiale o nome chimico CH-223191
Forma fisica Solid
CAS 301326-22-7
Colore Orange
Quantità 10 mg
Note sul grado di purezza Research
Formula molecolare C19H19N5O
Da utilizzare con (applicazione) Cancer-programmed cell death
Informazioni di solubilità DMSO : 125 mg/mL (374.94 mM; Need ultrasonic)
SMILES O=C(C1=CC=NN1C)NC2=CC=C(/N=N/C3=CC=CC=C3C)C=C2C
Molecular Weight (g/mol) 333.39
Peso formulazione 333.39
Percent Purity 98.03%
Grado Research
Vedi altri risultati Mostra meno risultati

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.