Learn More
Nabertherm Chamber Furnace LH 1200 Series with C440 Controller
Codice prodotto. p-7160785 Sfoglia Tutto Nabertherm Prodotti
Click to view available options
Capacità (sistema metrico):
15 L
30 L
60 L
120 L
216 L
Dimensione della confezione:
Pezzo
Codice prodotto. 15978330
Marca: Nabertherm LH0220NCNE
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Robust insulation of light refractory bricks. With a wide variety of optional equipment, these chamber furnaces can be optimally adapted to your processes.
  • Maximum Temperature: 1200 °C
  • Dual shell housing with rear ventilation, provides for low shell temperatures
  • Five-sided heating for very good temperature uniformity
  • Heating elements on support tubes ensure free heat radiation and a long service life
  • Controller mounted on furnace door and removable for comfortable operation
  • Protection of bottom heating and flat stacking surface provided by embedded SiC plate in the floor
  • Multi-layered, fiber-free insulation of light refractory bricks and special backup insulation
  • Door with brick-on-brick seal, hand fitted
  • Short heating times due to high installed power
  • Side vent with bypass connection for exhaust pipe
  • Self-supporting arch for high stability and greatest possible protection against dust
  • Quick lock on door
  • Freely adjustable air slide intake in furnace floor
  • Stand included
  • Defined application within the constraints of the operating instructions
Specifica
Capacità (sistema metrico) 15 L
Temperatura (sistema metrico) 1200°C
Controllo della temperatura C440
Dimensioni interne (L x P x A) 250 x 250 x 250 mm
Dimensioni esterne (L x P x A) 860 x 680 x 1215 mm
Peso (sistema metrico) 170 kg