missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Chart Recorder Paper

Codice prodotto. 13418696
Click to view available options
Temperature Range (Metric):
0°C to 50°C
Dimensione della confezione:
Pezzo
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 13418696

Marca: Eppendorf™ P06252111

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Includes chart recorder, RTD probe, keys, chart recorder paper and pens. Continuously monitor and record freezer temperature for up to seven days with this chart recorder.

  • Chart recorder paper
  • Type I
  • Front mounted to your freezer for easy access and can be fitted to all freezer models
  • Battery powered, will provide a minimum of eighteen months of operation
  • User configurable built in high and low temperature alarms
  • Can be factory installed with your freezer order at no additional charge or retrofitted on location
  • Selectable temperature range (0 to -50°C and -50 to -100°C) and rotation speed
TRUSTED_SUSTAINABILITY

Specifica

Temperature Range (Metric) 0°C to 50°C
Tipo Chart Recorder with Paper
Include Chart recorder, RTD probe, keys, chart recorder paper and pens
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.