Learn More
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Da utilizzare con (apparecchiatura):
™, Forma 900, Revco ExF/DxF, TSE/TSD Series
Forma 88300, HERAfreeze HFU300T, Revco UxF300
Forma 88400, TSU400
Forma 88500, Forma 88600, Forma 88700, TSU500, TSU600
Forma 900, HERAfreeze HFU-B, Revco ExF/DxF, TSE/TSD Series
HERAfreeze HFU-B, Forma 900, Revco ExF/DxF, TSE/TSD Series
HERAfreeze HFU700T, HERAfreeze HFU600T, Revco UxF500, Revco UxF600, Revco UxF700
Revco CxF 390, 350
Revco CxF Series (except ULT350 and ULT390)
Revco RDE, Forma FDE, TDE Series, HDE Series (500 box capacity models only)
Revco RDE, Forma FDE, TDE Series, HDE Series (600 box capacity models only)
Revco RLE, Forma 89000, TLE Series, HERAfreeze HLE 600
Revco RLE, Forma 89000, TLE Series, HLE Series (400 models only)
Revco RLE, Forma 89000, TLE Series, HLE Series (500 models only)
Serie Forma 900 e 7000, HERAfreeze HFU B, Revco ExF/DxF e TSE/TSD, Forma 900, Revco ExF/DxF, TSE/TSD Series
TDE Series Chest Freezers
TSE/TSD Series, Forma 900, HERAfreeze HFU-B, Revco ExF/DxF
TSU400, Revco UxF400
TSX Series
TSX400 Ultra-low Freezers
TSX500 Ultra-low Freezers
TSX600 Ultra-low Freezers
TSX700 Ultra-low Freezers
Thermo Scientific TSU Series, HERAfreeze HFU-T, Revco UxF
Tipo:
Carte per registratore diagrammi
Ink Chart Recorder, Factory Installed
Ink Chart Recorder, Field Installed
Inkless Chart Paper (pack of 50)
Inkless Chart Recorder, Factory Installed
Inkless Chart Recorder, Field Installed
Penna per registratore grafico
Dimensione della confezione:
Pezzo
pezzi 50
Descrizione
I registratori grafici per congelatori a bassissima temperatura Thermo Scientific™ consentono di registrare l'andamento settimanale delle temperature fino a -150°C. I registratori utilizzano carta per grafici 6 in. (15 cm) e possono essere installati in loco o in fabbrica.
Features:
- Seven-day recorder
- Field or factory installed
Ordering information:
- Field-installed units must be installed by a qualified professional
Specifica
Specifica
| Tipo | Inkless Chart Recorder, Field Installed |
| Gamma di temperatura | -115°C to +50°C |
| Da utilizzare con (apparecchiatura) | TSX Series |
| Autonomia fase statica | 7 Days |
| Opzioni di monitoraggio | Recorder |