missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA555689
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: AWWESNNYQS FCLPSEESEP EDLENGEESS AELDYQDPDA TSLKYVSGDV THPQAGAEDA LIVHCVDDSG HWGRGGLFTA LEKRSAEPR Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 80%.
In response to DNA strand breaks, chromatin adopts a relaxed structure due to the addition of poly(ADP-ribose) (PAR) to chromatin proteins by PARP enzymes (see PARP1; MIM 173870), and this relaxation facilitates the repair of DNA damage. CHD1L interacts with PAR and has a role in chromatin relaxation following DNA damage (Ahel et al., 2009 [PubMed 19661379]).
Specifica
| CHD1L | |
| Polyclonal | |
| Unconjugated | |
| CHD1L | |
| ALC1, CHDL, amplified in liver cancer 1, amplified in liver cancer protein 1, chromodomain-helicase-DNA-binding protein 1-like | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9557 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86WJ1 | |
| CHD1L | |
| Recombit protein corresponding to Human CHD1L | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto