Learn More
Aqualytic™ Checkit™ 5 in 1 Test Kit
Click to view available options
Quantità:
1 Pack
Dimensione della confezione:
Pezzo
Descrizione
Aqualytic™ Checkit™ 5 in 1 Test Kit includes all materials and operating instructions needed for mobile and static water analysis of 5 different components and comes in a sturdy and handy plastic case.
- Tests for chlorine, ph, cyanuric acid (turbidity method), calcium hardness (speed test), and total alkalinity (speed test)
- CHECKIT™ Comparator
- Compact, handy colorimetric unit suitable for mobile and stationary analyses
- Supplied with a generous number of different color scales
- CHECKIT™ Discs
- Continuous color scale makes it possible to achieve an exact color match between the standard and the sample.
- Specially manufactured in selected materials to remain color stable over a long period of time for reliable, reproducible measurement results
- AQUALYTIC™ tablet reagents (for 30 tests)
- Stirring Rod
- Cells
- Operating instructions
- Step-by-step explanation of how to conduct the test, ensuring reliable and fast and accurate measurements
Specifica
Specifica
| Include | Comparator apparatus, color comparison discs, cells, stirring rod, reagents, instructions |
| N. di test | 30 |
| Quantità | 1 Pack |
| Range | 6.5 to 8.4pH, 0 to 4.0mg/L Cl2, 20 to 200mg/L Cys, 20 to 800mg/L CaCO3, 20 to 800mg/L CaCO3 |
| Sufficiente per | 30 Tests |
| Metodo test | Turbidity, speed test |
| Tipo | Checkit™ 5 in 1 Test Kit |