Learn More
Lovibond™ Checkit™ Comparator Single Parameter Test Kit
Click to view available options
Range:
0 to 1mg/L
4 to 10pH
5.2 to 6.8mg/L
6 to 7.6mg/L
Dimensione della confezione:
Pezzo
Analiti:
Ammonia
Copper
pH
Descrizione
Aqualytic™ Checkit™ Comparator Single Parameter Test Kit includes all materials and operating instructions needed for mobile and static water analysis of a single parameter and comes in a sturdy and handy plastic case.
- Compact and lightweight
- Provides the basis for a comprehensive and easy-to-use analysis system
- Comparator Discs contain a continuous color scale for exact color matching between standard and sample
Specifica
Specifica
| Include | Comparator apparatus, color comparison discs, cells, stirring rod, reagents, instructions |
| N. di test | 30 |
| Quantità | 1 Pack |
| Range | 4 to 10pH |
| Tipo | Checkit™ Comparator Single Parameter Test Kit |