Descrizione
- Made from strong 92 GSM non-microporous PE laminate for durability and protection.
- Double front closing system and large zip pull ensure easy wear and secure fit.
- Provides excellent barrier protection against liquid splashes, jet sprays (Type 3), and hazardous particles (TIL Class 2, EN 1073-2).
- Meets multiple standards: EN 14126, EN 1073-2, EN 1149, EN ISO 13688, Type 6 (EN 13034), Type 5 (EN ISO 13982-1), Type 4 and Type 3 (EN 14605).
- Gray color design for low-visibility and discreet applications such as first responders.
- Chemically resistant taped seams provide enhanced protection and strength.
- High chemical permeation resistance tested against multiple chemicals.
- Passes all EN 14126 tests, achieving the highest protection class.
- Offers high particle holdout (TIL Class 2, EN 1073-2).
- Reliable safety and protection for the workforce.
Specifica
Specifica
| Product Type | Protective Coveralls |
| Size | Small |
| Sesso | Unisex |
| Materiale | Polyethylene, Polypropylene |
| Colore | Gray |
| Monouso | Single-use |
| Stile vita | Standard |
| Stile della caviglia | Open |
| Stile polso | Open |
| Stile collo | Hooded |
| Vedi altri risultati |