Learn More
Thermo Scientific™ Cheminert™ Sample injector loops, PAEK, 50μL
Click to view available options
Volume (sistema metrico) di campione:
50 μL
Dimensione della confezione:
Pezzo
Descrizione
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves. It can be actuated manually or with a microelectronic actuator. With 1/16 in. fittings and 0.010 in. ports, it is available in six-port or ten-port configurations and includes a 5μL sample loop.
- 1/16″ fittings
- 0.010″ ports
- Available in 6-port or 10-port configurations
- Available with manual or microelectric actuator
Specifica
Specifica
| Volume (sistema metrico) di campione | 50 μL |
| Tipo | Sample Injector Loop |
| Materiale statore | PEEK |