Learn More
Saint Gobain Life Sciences Chemware™ PFA Jar with Cap
Click to view available options
Capacità (sistema metrico):
120 mL
360 mL
480 mL
1000 mL
2000 mL
Dimensione della confezione:
Pezzo
Descrizione
- Chemware™ PFA jars store solutions, paste and bulk materials with no concern about leaching, corrosion, contamination or evaporation.
- They are resistant to corrosive liquids, acids, solvents, sensitive reagents, and staining solutions.
- PFA jars feature a large mouth for easy access and non-stick surface for easy cleaning.
- There is an area for identification marking on side of the jars.
- Stackable, these jars have a knurled top for secure grip to facilitate easy opening and closing.
Specifica
Specifica
| Capacità (sistema metrico) | 120 mL |
| Diametro (sistema imperiale) | 8365008 |
| Materiale | PFA |
| Closure Included | Sì |
| Bocca | Larga |
| Altezza (sistema metrico) | 53.2 mm |
| Product Type | Jar |
| Altezza (sistema imperiale) | 2.1 in. |
| Materiale chiusura | PFA |
| Diametro (sistema metrico) | 66 mm |