missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken anti-Mouse IgG (H+L) Secondary Antibody, HRP (Pre-adsorbed), Novus Biologicals™
Chicken Polyclonal Antibody
Marca: Novus Biologicals NBP1-75109
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifica
| IgG (H+L) | |
| Polyclonal | |
| HRP | |
| Chicken | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Rehydrate with 0.55 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. | |
| Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:200-1:5000, ELISA 1:200-1:5000, Immunohistochemistry 1:500-1:5000, Immunohistochemistry-Paraffin | |
| Purified Mouse IgG (H&L) | |
| 0.5 mg | |
| Adaptive Immunity, Immunology | |
| Based on IEP, this antibody reacts with heavy gamma chains on mouse IgG and light chains on all mouse immunoglobulins. This antibody has been pre-adsorbed against human and rabbit IgG/serum proteins. | |
| Mouse | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto