missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken anti-Rabbit IgG (H+L) Secondary Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
131.00€
Specifica
| Antigene | IgG (H+L) |
|---|---|
| Concentrazione | 4.5 mg/mL |
| Applicazioni | Immunoassay |
| Classificazione | Polyclonal |
| Coniugato | Unconjugated |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
18536172
|
Novus Biologicals
NBP1-75258-0.5mg |
0.5 mg |
131.00€
0.50mg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay.Specifications
| IgG (H+L) | |
| Immunoassay | |
| Unconjugated | |
| RUO | |
| Rabbit | |
| IgG | |
| Affinity purified | |
| Based on IEP, this antibody reacts with heavy gamma chains on rabbit IgG and light chains on all rabbit immunoglobulins |
| 4.5 mg/mL | |
| Polyclonal | |
| Chicken | |
| Adaptive Immunity, Immunology | |
| Purified Rabbit IgG (H&L) | |
| Secondary | |
| Store at 4°C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title