Learn More
Thermo Scientific™ Chiller Plumbing Kits and Accessories
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Tipo:
Fittings
Plumbing Package
Tubing
Tubing Kit
Da utilizzare con (apparecchiatura):
Merlin Chillers with PD1/PD2 pump
ThermoFlex 3500 to 5000
ThermoFlex 900 to 10000
ThermoTemp, Polar
ThermoTemp, Polar, VersaCool
Size:
1/2 in. NPT, 25 ft.
1/2 in. NPT, 50 ft.
1/2 to 3/8 in. NPT
3/4 in. NPT, 32.8 ft.
6 mm ID per meter, 0.5 meter long
6 mm ID per meter, 1.0 m
6 mm ID per meter, 1.0 meter long
6 mm ID per meter, 1.5 m
6 mm ID per meter, 1.5 meter long
8 mm ID per meter
Materiale:
Brass
Flexible Plastic
Perbunan
Polybraid
Stainless-steel
Dimensione della confezione:
1m
Pezzo
pezzi 2
Descrizione
Thermo Scientific™ Chiller Plumbing Kits and accessories include flexible tubing, tubing insulation, and hose clamps.
Features:
- Include flexible tubing, tubing insulation, and hose clamps
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | ThermoTemp, Polar, VersaCool |
| Tipo | Tubing |
| Include | Insulated flexible SS tubing with M16x1 threaded fittings 0.5 meter long |
| Materiale | Stainless-steel |
| Size | 6 mm ID per meter, 0.5 meter long |