Learn More
CHL-1/L1CAM-2 Rat anti-Human, Clone: 316223, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Marca: R&D Systems MAB2126-SP
Descrizione
CHL-1/L1CAM-2 Monoclonal specifically detects CHL-1/L1CAM-2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifica
| CHL-1/L1CAM-2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 μg/mL, Immunohistochemistry 1.7-25 μg/mL | |
| NP_006605 | |
| CHL1 | |
| Mouse myeloma cell line NS0-derived recombinant human CHL-1/L1CAM-2 Ile25-Gln1096 Accession # NP_006605 | |
| 25 μg | |
| Primary | |
| Detects human CHL-1/L1CAM-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CHL-1 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 316223 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CALL, CALLClose homolog of L1, cell adhesion molecule with homology to L1CAM (close homolog of L1), cell adhesion molecule with homology to L1CAM (close homologue of L1), CHL1, FLJ44930, L1CAM-2, L1CAM2L1 cell adhesion molecule 2, MGC132578, neural cell adhesion molecule L1-like protein | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10752 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.