Learn More
Macherey-Nagel™ Cartuccia HR-P Chromafix™
Sfoglia Tutto Macherey-Nagel Prodotti
Click to view available options
Peso del letto:
200 mg
330 mg
680 mg
Dimensione della confezione:
pezzi 50
Descrizione
Features
- Highly porous polystyrene-divinylbenzene copolymer
- Specific surface 1200 m2/g
- Particle size 50–100 μm
- Very high binding capacity, up to 30 percent of adsorbent weight (for comparison: silica adsorbents about 3 percent)
Recommended application:
- Aromatic compounds
- Phenols from water
- Nitroaromatics from water
- Pesticides from water
- PAHs from oil
Specifica
Specifica
| Peso del letto | 680 mg |
| Da utilizzare con (applicazione) | Aromatic Compounds, Phenols From Water, Nitroaromatics From Water, Pesticides From Water, PAHs From Oil |
| Dimensione particelle | 50 to 100 μm |
| Assorbente | HR-P |
| Tipo | Chromafix HR-P Cartridge |
| Quantità | 50 Pack |
| Composizione resina | Polystyrene-divinylbenzene adsorbent resin |
| Area della superficie (sistema metrico) | 1200 m2/g |