Learn More
Proteintech ChromoTek Binding Control Agarose
Codice prodotto. 17333283
Click to view available options
Quantità:
20 Reactions
Dimensione della confezione:
20 reazioni
Codice prodotto. 17333283
Marca: Proteintech BAB20
Control agarose resins that consist of the same agarose matrix as our Nano-Traps Agarose.
  • Control of unspecific binding of proteins, DNA, etc. to agarose beads
  • Pre-clearing of cell lysate
  • 20 reactions / 500 μL slurry
  • Stored in 20% EtOH
Specifica
Quantità 20 Reactions
Sufficiente per 20 Reactions