Learn More
Proteintech ChromoTek iST mNeonGreen-Trap Kit
Codice prodotto. 17363393
Click to view available options
Quantità:
8 Reactions
Dimensione della confezione:
8 reazioni
Codice prodotto. 17363393
Marca: Proteintech ntakiST8
Kit for AP-MS sample preparation of mNeonGreen-fusion proteins. Includes 8 reactions plus 2 controls.
  • Kit for IP/ Co-IP of mNeonGreen tagged proteins and sample preparation for mass spectrometry (MS)
Specifica
Quantità 8 Reactions
Sufficiente per 8 Reactions