Learn More
Proteintech ChromoTek Mdm4/ HdmX-Trap Agarose
Click to view available options
Sufficiente per:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Dimensione della confezione:
10 reazioni
100 reazioni
20 reazioni
200 reazioni
400 reazioni
Description
- Coupled VHH: Recombinant, monoclonal anti-Mdm4/HdmX single domain antibody (sdAb) fragment
Specifications
Specifications
| Da utilizzare con (applicazione) | IP, CoIP |
| Bersaglio | Double minute 4 protein (Mdm4/HdmX), human. Note that binding of Mdm4/HdmX-Trap interferes with binding of p53 to Mdm4 HdmX. |
| Label or Dye | Agarose |
| Specie | Alpaca |
| Sufficiente per | 20 Reactions |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction