Learn More
Proteintech ChromoTek PARP1-Trap Agarose Beads Kit
Click to view available options
Quantità:
20 Reactions
Dimensione della confezione:
20 reazioni
Descrizione
- PARP1-Trap Agarose is an affinity reagent for IP of human PARP1
- Consists of a PARP1 Nanobody/ VHH, which is coupled to agarose beads
Specifica
Specifica
| Quantità | 20 Reactions |
| Sufficiente per | 20 Reactions |