Learn More
Proteintech ChromoTek Spot-Trap™ Agarose Kit
Click to view available options
Quantità:
20 Reactions
Dimensione della confezione:
20 reazioni
Descrizione
- Spot-Trap™ Agarose is an affinity bead for immunoprecipitation (IP) of Spot-tagged proteins
- Comprises an anti-Spot-Tag™ VHH/ Nanobody coupled to agarose beads
Specifica
Specifica
| Quantità | 20 Reactions |
| Sufficiente per | 20 Reactions |