Learn More
CHTF18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13030-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CHTF18, CHTF8.Specifica
| CHTF18 | |
| Polyclonal | |
| Unconjugated | |
| CHTF18 | |
| C16orf41, C321D2.2, C321D2.3, C321D2.4, CHL12, CHTF18, Ctf18, hCTF18, RUVBL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 214901, 287146, 63922 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BIW9, Q8WVB6 | |
| Chtf18 | |
| CHTF18 Fusion Protein Ag3672 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |