Learn More
Masterflex™ Cigarette Lighter Adapter Cable
Click to view available options
Overall Length (English):
25 ft.
Dimensione della confezione:
Pezzo
Descrizione
- Choose accessories and replacement parts suitable for your Masterflex™ pump
- Power your pump on-the-go
Specifica
Specifica
| Descrizione | Cigarette lighter adapter cable, 25 ft |
| Overall Length (English) | 25 ft. |
| Tipo | Cigarette Lighter Adapter |