missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cimarec™ i Bath Mounts

Codice prodotto. p-4531290 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Materiale:
Acrylic
Stainless Steel
Dimensioni (L x P x A):
520 x 315 x 160 mm
610 x 330 x 185 mm
Dimensione della confezione:
Pezzo
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 10341262

Marca: Thermo Scientific™ 50087880

Solo null disponibili
Add to basket
Add to basket
This item is not returnable. View return policy

Use Thermo Scientific™ Cimarec™ i Bath Mounts with Telesystem series stirrers.

Use Thermo Scientific™ Cimarec™ i Bath Mounts with Telesystem series stirrers.

  • Available in stainless steel and transparent acrylic
  • Capacity: 3.9 gal. (15L)
TRUSTED_SUSTAINABILITY

Specifications

Da utilizzare con (apparecchiatura) Telesystem Series Stirrer
Tipo Bath Mount
Dimensioni (L x P x A) 610 x 330 x 185 mm
Materiale Acrylic
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.