Learn More
Ciprofibrate, LoGiCal
Codice prodotto. 30458180
Click to view available options
Quantità:
10 mg
Dimensione della confezione:
10mg
Codice prodotto. 30458180
Marca: LoGiCal LGCFOR1141.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Ciprofibrate
CAS 52214-84-3
Nomi componenti o analita Ciprofibrate
Shipping Condition Room Temperature
Formula molecolare C13 H14 Cl2 O3
Certificazione/Conformità ISO 9001
Sinonimo Ciprofibrate, 2-[4-[(1RS)-2,2-Dichlorocyclopropyl]phenoxy]-2-methylpropanoic acid
InChI Formula InChI=1S/C13H14Cl2O3/c1-12(2,11(16)17)18-9-5-3-8(4-6-9)10-7-13(10,14)15/h3-6,10H,7H2,1-2H3,(H,16,17)
Tipo di soluzione Neat
SMILES CC(C)(Oc1ccc(cc1)C2CC2(Cl)Cl)C(=O)O
IUPAC Name 2-[4-(2,2-dichlorocyclopropyl)phenoxy]-2-methylpropanoic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 289.15
Quantità 10 mg
Peso formulazione 288.032 g/mol