Learn More
Citric Acid tert-Butyl Ester, TRC
Codice prodotto. 30362052
Cambia vista
Click to view available options
Quantità:
25 mg
250 mg
Dimensione della confezione:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30362052 25 mg 25mg
30357086 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 30362052 Fornitore Toronto Research Chemicals N. del fornitore TRCC52100525MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 114340-52-2
Formula molecolare C10H16O7
Molecular Weight (g/mol) 248.23
Sinonimo 3-tert-Butyl Citrate, 2-Hydroxy-1,2,3-propanetricarboxylic Acid 2-(1,1-Dimethylethyl) Ester
IUPAC Name 3-hydroxy-3-[(2-methylpropan-2-yl)oxycarbonyl]pentanedioic acid
SMILES CC(C)(C)OC(=O)C(O)(CC(=O)O)CC(=O)O
Specifica
Materiale o nome chimico Citric Acid tert-Butyl Ester
Quantità 25 mg
InChI Formula InChI=1S/C10H16O7/c1-9(2,3)17-8(15)10(16,4-6(11)12)5-7(13)14/h16H,4-5H2,1-3H3,(H,11,12)(H,13,14)
Peso formulazione 248.0896