Learn More
Heidolph™ Morsetto: per matracci di Erlenmeyer
Sfoglia Tutto Heidolph Instruments Prodotti
Click to view available options
Capacità:
100 mL Erlenmeyer flasks
1000 mL Erlenmeyer flasks
2000 mL Erlenmeyer flasks
25 mL Erlenmeyer flasks
250 mL Erlenmeyer flasks
50 mL Erlenmeyer flasks
500 mL Erlenmeyer flasks
Dimensione della confezione:
Pezzo
Size:
6
Da utilizzare con (apparecchiatura):
Perforated platforms 100, 1000 and 2000
Descrizione
- Maximum number of clamps per platform type:
- 549-51000-00
- For platform 100 - max. 16
- For platform 1000 - max. 20
- For platform 2000 - max. 36
- 549-52000-00
- For platform 100 - max. 16
- For platform 1000 - max. 20
- For platform 2000 - max. 36
- 549-55000-00
- For platform 100 - max. 3
- For platform 1000 - max. 4
- For platform 2000 - max. 9
- 549-56000-00
- For platform 100 - max. 2
- For platform 1000 - max. 4
- For platform 2000 - max. 5
- 549-63000-00
- For platform 1000 - max. 2
- For platform 2000 - max. 3
- 549-51000-00
Specifica
Specifica
| Capacità | 25 mL Erlenmeyer flasks |
| Product Type | Clamp |
| Da utilizzare con (apparecchiatura) | Perforated platforms 100, 1000 and 2000 |
| Size | 1 |