Learn More
Thermo Scientific™ Clamps for CO2 Resistant Shaker
Codice prodotto. 15371115 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Product Type:
Clip
Flask Clamp
Funnel Clamp
Infusion bottle clamp
Starter Kit
Morsetto:
100 mL
1000 mL
150 mL
200 mL
2000 mL
25 mL
250 mL
3000 mL
50 mL
500 mL
5000 mL
Flask
Separatory
Dimensione della confezione:
1 unità
Pezzo
Codice prodotto. 15371115
Marca: Thermo Scientific™ 88881116
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Achieve maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
Maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
  • Compatible with Thermo Scientific™ CO2Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
  • Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.
TRUSTED_SUSTAINABILITY
Specifica
Product Type Infusion bottle clamp
Morsetto 1000 mL
Da utilizzare con (apparecchiatura) CO2 Resistant Shaker
N. di posizioni 6