Learn More
Zwiebel™ Class F1 Box Weights
Click to view available options
Massa:
50 mg
200 mg
500 mg
2 g
50 g
100 g
200 g
1000 g
2000 g
Tolleranza:
±0.04 mg
±0.06 mg
±0.08 mg
±0.12 mg
±0.30 mg
±0.50 mg
±1.00 mg
±10 mg
±5.00 mg
Dimensione della confezione:
Pezzo
Description
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
| Massa | 50 mg |
| Tolleranza | ±0.04 mg |
| Da utilizzare con (applicazione) | For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II |