Learn More
Zwiebel™ Class M2 Weight Set
Click to view available options
Massa:
1 to 100 g
1 to 200 g
1 to 50 g
1 to 500 g
Dimensione della confezione:
Pezzo
Descrizione
- Class M2 stainless steel calibration weight
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifica
Specifica
| Massa | 1 to 50 g |
| Da utilizzare con (applicazione) | For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II |
Titolo del prodotto
Spot an opportunity for improvement?Share a Content Correction