Learn More
Abeba™ Classic 31000 Shoes
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
Dimensione della confezione:
1 paio
Descrizione
- CE EN ISO 20345:2011 SB SRC
- Grained leather
- White
- Steel toe cap
- Slip resistance PU outsole sole
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Lining microfiber, respiration absorbing
- Adjustable instep area
- Buckle fastening
- Heel patch microfiber
Specifica
Specifica
| Certificazione/Conformità | CE EN ISO 20345:2011 SB SRC |
| Colore | White |
| Materiale rivestimento interno | Microfiber |
| Materiale | Leather |
| Size | 36 |
| Caratteristiche suola | Slip-resistant |
| Materiale suola | Polyurethane |
| Stile | 31000 |
| Materiale dello zoccolo | Steel |
| Resistente a | Animal fats, oil and fuel |