Learn More
Abeba™ Classic 31001 Shoes
Click to view available options
Size:
36
37
38
39
40
41
42
43
45
46
Dimensione della confezione:
1 paio
Descrizione
- CE EN ISO 20345:2011 SB A, E, FO SRC
- Microfiber
- White
- Steel toe cap
- Slip resistance PU outsole sole
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Microfiber lining and heel patch
- Adjustable instep area
- Buckle fastening
- Comfortable shape
Specifica
Specifica
| Certificazione/Conformità | CE EN ISO 20345:2011 SB A, E, FO SRC |
| Colore | White |
| Materiale rivestimento interno | Microfiber |
| Materiale | Microfiber |
| Size | 46 |
| Caratteristiche suola | Slip-resistant |
| Materiale suola | Polyurethane |
| Stile | 31001 |
| Materiale dello zoccolo | Steel |
| Resistente a | Animal fats, oil and fuel |