missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CLDN3 (Human) Recombinant Protein
Human CLDN3 full-length ORF ( AAH16056, 1 a.a. - 220 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00001365-P01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
- Sequence: MSMGLEITGTALAVLGWLGTIVCCALPMWRVSAFIGSNIITSQNIWEGLWMNCVVQSTGQMQCKVYDSLLALPQDLQAARALIVVAILLAAFGLLVALVGAQCTNCVQDDTAKAKITIVAGVLFLLAALLTLVPVSWSANTIIRDFYNPVVPEAQKREMGAGLYVGWAAAALQLLGGALLCCSCPPREKKYTATKVVYSAPRSTGPGASLGTGYDRKDYV
Specifica
AAH16056 | |
claudin 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLDN3 | |
GST |
1365 | |
Wheat germ expression system | |
10 μg | |
C7orf1, CPE-R2, CPETR2, HRVP1, RVP1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |