Learn More
Sampling Systems™ Cleaning Coupons for Metal
Sfoglia Tutto Sampling Systems Prodotti
Click to view available options
Materiale:
Aluminum
Stainless Steel (304)
Stainless Steel (316)
Dimensioni (L x P):
100 x 100 mm
50 x 50 mm
50 x 50 mm
Finitura:
Bright Polish
Satin finish
Dimensione della confezione:
Pezzo
Descrizione
- Tests available for aluminum, 304 stainless steel and 316 stainless steel
- All metals also available for either bright polish or satin finish
- Two sizes available for each configuration, either 50 x 50mm or 100 x 100mm
Specifica
Specifica
| Product Type | Cleaning Coupon |
| Dimensioni (L x P) | 50 x 50 mm |
| Finitura | Satin finish |
| Da utilizzare con (applicazione) | Testing and Validating Cleaning Regime |
| Materiale | Stainless Steel (304) |