Learn More
Cole-Parmer Cleanroom Female Luer Fittings, Straight Union
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Materiale:
CrystalVu (CVU)
Kynar
Nylon
Polypropylene
Colore:
Clear
Natural
White
Dimensione della confezione:
pezzi 10
Descrizione
Forms a leak-free luer connection with a quick simple turn. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application
Specifica
Specifica
| Certificazione/Conformità | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS |
| Materiale | Polypropylene |
| Quantità confezione | 10/Pk. |
| Ingresso | Female Luer |
| Colore | Clear |