Learn More
Cole-Parmer Cleanroom Female Luer to Hose Barb Fittings, Straight Adapter, White Nylon
Codice prodotto. p-200105076 Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Diametro (sistema imperiale) interno:
1/16 in.
1/4 in.
1/8 in.
3/16 in.
3/32 in.
5/32 in.
Dimensione della confezione:
pezzi 10
Codice prodotto. 30054235
Marca: Cole-Parmer 5010987
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Forms a leak-free luer connection with a quick simple turn. Single barb design tightly grips the tubing. Meets FDA regulations 21 CFR 178.3297 and RoHS compliant
Specifica
Certificazione/Conformità FDA;Non-DEHP;Non-Phthalate;RoHS
Diametro (sistema imperiale) interno 3/16 in.
Materiale Nylon
Quantità confezione 10/Pk.
Ingresso Female Luer x Hose Barb
Colore White
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.