Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Straight Reducer, White Nylon
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Dimensioni dei tubi:
1/4 to 1/8 in.
1/4 to 3/16 in.
1/4 to 5/32 in.
1/8 to 1/16 in.
1/8 to 3/32 in.
3/16 to 1/8 in.
3/16 to 3/32 in.
3/32 to 1/16 in.
3/8 to 1/4 in.
5/16 to 1/4 in.
5/32 to 1/8 in.
5/32 to 3/32 in.
Dimensione della confezione:
pezzi 10
Descrizione
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. FDA and RoHS compliant
Specifica
Specifica
| Certificazione/Conformità | FDA;Non-DEHP;Non-Phthalate;RoHS |
| Quantità confezione | 10/Pk. |
| Dimensioni dei tubi | 1/8 to 3/32 in. |
| Porta di uscita | Hose Barb Reducer |
| Metodo di sterilizzazione | E-beam/gamma irradiation, EtO, or autoclave up to 121°C |
| Forma | Diritto |
| Colore | White |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.