Learn More
Cole-Parmer Cleanroom Large-Bore Female Luer to Hose Barb Fittings, Straight Adapter, White Nylon
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Diametro (sistema imperiale) interno:
1/4 in.
3/16 in.
5/16 in.
Dimensione della confezione:
pezzi 10
Descrizione
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Large-bore luer fittings offer higher flow rates compared to standard luers. Single barb design tightly grips the tubing. Meets FDA regulations 21 CFR 178.3297 and RoHS compliant
Specifica
Specifica
| Certificazione/Conformità | FDA;Non-DEHP;Non-Phthalate;RoHS |
| Diametro (sistema imperiale) interno | 3/16 in. |
| Materiale | Nylon |
| Quantità confezione | 10/Pk. |
| Ingresso | Large Bore Female Luer, Hose Barb |
| Colore | White |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction