Learn More
Cole-Parmer Cleanroom Large-Bore Male Luer to Hose Barb Fittings, Straight Adapter, Natural Nylon
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Diametro (sistema imperiale) interno:
1/4 in.
3/16 in.
5/16 in.
Dimensione della confezione:
pezzi 10
Descrizione
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Large-bore luer fittings offer higher flow rates compared to standard luers. Single barb design tightly grips the tubing. RoHS compliant
Specifica
Specifica
| Certificazione/Conformità | Non-DEHP;Non-Phthalate;RoHS |
| Diametro (sistema imperiale) interno | 5/16 in. |
| Materiale | Nylon |
| Quantità confezione | 10/Pk. |
| Ingresso | Large Bore Male Luer, Hose Barb |
| Colore | Natural |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.