Learn More
Cole-Parmer Cleanroom Male Luer Lock Plug and Nut Assemblies, Straight Adapter, White Nylon
Codice prodotto. 30054317 Sfoglia Tutto Cole-Parmer Prodotti
Cambia vista
Click to view available options
Colore:
Berry
Lime
Natural
Orange
Rose
Violet
Yellow
Dimensione della confezione:
pezzi 10
Codice prodotto. Colore unitSize
30054317 Berry pezzi 10
30053703 Yellow pezzi 10
30052930 Violet pezzi 10
30054267 Natural pezzi 10
30054012 Orange pezzi 10
30053245 Lime pezzi 10
30053675 Rose pezzi 10
7 options
Codice prodotto. 30054317
Marca: Cole-Parmer 5011042
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
This item is not returnable. View return policy
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Nut and body rotate independently during connection
Specifications
Certificazione/Conformità FDA;Non-DEHP;Non-Phthalate;RoHS
Materiale Nylon
Ingresso Male Luer Lock, Plug
Colore Berry
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.