Learn More
Cole-Parmer Cleanroom Panel-Mount Female Luer to Hose Barb Fittings, Straight Adapter, Animal-Derivative-Free Polypropylene
Codice prodotto. 30054141 Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Diametro (sistema imperiale) interno:
1/16 in.
1/8 in.
3/32 in.
Dimensione della confezione:
pezzi 10
Codice prodotto. 30054141
Marca: Cole-Parmer 5011431
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Forms a leak-free luer connection with a quick simple turn. Single barb design tightly grips the tubing. Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Specifica
Certificazione/Conformità ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS
Diametro (sistema imperiale) interno 3/32 in.
Materiale Polypropylene
Quantità confezione 10/Pk.
Ingresso Female Luer, Hose Barb
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.