Learn More
Cole-Parmer Cleanroom Stationary Male Luer Lock to Barb Fittings, Straight Adapter, Animal-Derivative-Free Polypropylene
Codice prodotto. p-200105049 Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Diametro (sistema imperiale) interno:
1/16 in.
1/4 in.
1/8 in.
3/16 in.
3/32 in.
5/32 in.
Dimensione della confezione:
pezzi 10
Codice prodotto. 30053949
Marca: Cole-Parmer 5011468
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Connect to a female luer body to form a leak-free fluid path. Single barb design tightly grips the tubing. Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Specifica
Certificazione/Conformità ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS
Diametro (sistema imperiale) interno 5/32 in.
Materiale Polypropylene
Quantità confezione 10/Pk.
Ingresso Male Luer x Hose Barb