Learn More
Cole-Parmer Cleanroom Stationary Male Luer Lock to Barb Fittings, Straight Adapter, CrystalVu™
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Diametro (sistema imperiale) interno:
1/16 in.
1/4 in.
1/8 in.
3/16 in.
3/32 in.
5/32 in.
Dimensione della confezione:
pezzi 10
Descrizione
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Connect to a female luer body to form a leak-free fluid path. Single barb design tightly grips the tubing. USP Class VI, EU 2002/72/EC, and RoHS compliant
Specifica
Specifica
| Certificazione/Conformità | Non-DEHP;Non-Phthalate;RoHS;USP |
| Diametro (sistema imperiale) interno | 3/16 in. |
| Quantità confezione | 10/Pk. |
| Ingresso | Male Luer x Hose Barb |