Learn More
Micronova™ Cleanroom Vinyl Tape
Codice prodotto. p-200136338 Sfoglia Tutto Micronova Prodotti
Click to view available options
Colore:
Dark Blue
Light Blue
Light Green
Red
Yellow
Dimensione della confezione:
pezzi 24
pezzi 36
pezzi 48
Codice prodotto. 30694883
Marca: Micronova™ CR100PC1LB
Made with rubber, clean-peel adhesive is extremely conformable, making it ideal for color coding, marking and sealing curved and irregular surfaces
  • The Cleanroom Vinyl Tape offers a clean peel from wafer cartridges and cleanroom work surfaces and the matte finish can be written on with a cleanroom pen
  • Special processing and packaging techniques ensure low particles and extractable ion counts for applications in and around the cleanroom
  • Processed for applications in and around cleanrooms
  • Vinyl tape with synthetic rubber adhesive
  • Ideal for taping and sealing curved and irregular surfaces
  • Matte finish can be written on with a cleanroom pen
  • Specially processed and packaged for the cleanroom
  • Available in a variety of colors and widths
  • Individually packaged in a polyethylene bag
  • 36 yards wound on a 3 in. plastic core
  • Contact your Fisher Safety Customer Service Representative at 1-800-772-6733 for ordering information on colors not shown here
Specifica
Autoclavabile Autoclavable
Tipo di adesivo Rubber
Rivestimento Vinyl
Colore Light Blue
Finitura Matte
Formato Roll
Materiale Vinyl
Product Type Cleanroom Tape
Sterilità Non-sterile
Senza lattice No
Dimensions (LxW) 329.1 x 2.54 cm
Lunghezza (sistema imperiale) 108 ft.
Lunghezza (sistema metrico) 329.1 cm
Larghezza (sistema imperiale) 1 in.
Larghezza (sistema metrico) 2.54 cm