Learn More
Thermo Scientific™ Closures with Prefitted Septa for 13mm Crimp/Snap-Ring LoVials™
Click to view available options
Setto:
Red PTFE/White Silicone
Dimensione della confezione:
pezzi 144
Descrizione
Thermo Scientific™ National Closures with Prefitted Septa are available in easy-to-apply Kim-Snap or standard aluminum variety to suit your application needs.
- Kim-Snap™ closures provide a tight seal without the need of a crimper
- National Scientific aluminum crimp closures provide a secure, leak-resistant seal
- Aluminum seals must be applied with a crimping tool
Specifica
Specifica
| Materiale | PTFE, Silicone |
| Setto | Red PTFE/White Silicone |
| Dimensione chiusura | 13 mm |