Learn More
Tocris Bioscience™ CLP 257
Click to view available options
Quantità:
10 mg
50 mg
Dimensione della confezione:
10mg
50mg
Descrizione
CLP 257 is a selective KCC2 K+-Cl- cotransporter activator (EC50 = 616 nM). Exhibits selectivity for KCC2 over other KCC family members, NKCC1 and GABAA receptors, as well as 55 other receptors. Enhances Cl- transport and increases KCC2 plasma membrane expression in spinal slices with reduced KCC2 function. Alleviates hypersensitivity in rats with peripheral nerve injury.
Specifica
Specifica
| Materiale o nome chimico | (5Z)-5-[(4-Fluoro-2-hydroxyphenyl)methylene]-2-(tetrahydro-1-(2H)-pyridazinyl)-4(5H)-thiazolone |
| CAS | 1181081-71-9 |
| Quantità | 50 mg |
| Bersaglio | KCC2 Activators |
| Formula molecolare | C14H14FN3O2S |
| Peso formulazione | 307.34 |
| Purezza | >98% |